Silicon standard solution, 10000 mg/L Si in water (from Si) standard for AAS and ICP
Catalog # JOBIPLSI9A-3X
Supplier: JOBIN YVON EMISSION
CAS Number:
Specifications
- Concentration:10000 mg/L
- Matrix:H₂O
- Pack type:Glass bottle
- Pk:500 mL
- Element:Silicon
- Symbol:Si
Specifications
About this item
Certifications: Traceable to SRM from NIST