Scandium standard solution, 1000 mg/L Sc in 2 M nitric acid (from Sc₂O₃) standard for AAS
Catalog # KRAF03772.2000
Supplier: Bernd Kraft
CAS Number:
Specifications
- Concentration:1000 mg/L
- Matrix:2 M HNO₃
- Pack type:Plastic bottle
- Pk:500 mL
- Element:Scandium
- Symbol:Sc
Specifications
About this item
Certifications: Traceable to NIST