Rubidium standard solution, 1000 mg/L Rb in 0,5 M nitric acid (from Rb₂CO₃) standard for ICP
Catalog # KRAF03737.0000
Supplier: Bernd Kraft
CAS Number:
Specifications
- Concentration:1000 mg/L
- Matrix:0,5 M HNO₃
- Pack type:Plastic bottle
- Pk:100 mL
- Element:Rubidium
- Symbol:Rb
Specifications
About this item
Certifications: Traceable to NIST