Neodymium standard solution, 1000 mg/L Nd in 0,5 M nitric acid (from Nd₂O₃) standard for ICP
KRAF03730.0000
:
- Concentration:1000 mg/L
- Matrix:0,5 M HNO₃
- Pack type:Plastic bottle
- Pk:100 mL
- Element:Neodymium
- Symbol:Nd
Traceable to NIST
: Traceable to NIST