Pesticide reference standard, Clofenotane 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Catalog # EHERL12082000CY
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:50-29-3
- Description:Clofenotane 10 µg/ml in cyclohexane
- Formula:C₁₄H₉Cl₅
- Description:Clofenotane