Vanadium standard solution, 5000 mg/L V in 4 M nitric acid (from (NH₄)VO₃) standard for AAS
Catalog # KRAF03661.2000
Supplier: Bernd Kraft
CAS Number:
Specifications
- Concentration:5000 mg/L
- Matrix:4 M HNO₃
- Pack type:Plastic bottle
- Pk:500 mL
- Element:Vanadium
- Symbol:V
Specifications
About this item
Certifications: Traceable to NIST