Order Entry
Germany
ContactUsLinkComponent
Praseodymium standard solution, 1000 mg/L Pr in 0,5 M nitric acid (from Pr₆O₁₁) standard for ICP
Praseodymium standard solution, 1000 mg/L Pr in 0,5 M nitric acid (from Pr₆O₁₁) standard for ICP
Catalog # KRAF03734.0000
Supplier:  Bernd Kraft
CAS Number:  
Praseodymium standard solution, 1000 mg/L Pr in 0,5 M nitric acid (from Pr₆O₁₁) standard for ICP
Catalog # KRAF03734.0000
Supplier:  Bernd Kraft
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    0,5 M HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Element:
    Praseodymium
  • Symbol:
    Pr

Specifications

About this item

Certifications: Traceable to NIST