Order Entry
Germany
ContactUsLinkComponent
1,2,3,4-Tetrahydroisoquinoline
1,2,3,4-Tetrahydroisoquinoline
Catalog # MOLEM86277908
Supplier:  Molekula
CAS Number:  
1,2,3,4-Tetrahydroisoquinoline
Catalog # MOLEM86277908
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :133.19 g/mol
EINECS:202-050-0
CAS Number:91-21-4
Formula:C₉H₁₁N
MDL Number:MFCD00006896