1,2,3,4-Tetrahydroisoquinoline ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:91-21-4
:
Formula:C₉H₁₁N
MDL Number:MFCD00006896
:133.19 g/mol
Boiling Pt:232…233 °C
Melting Pt:–30 °C
Density:1.049
Flash Pt:98 °C (208 °F)
EINECS:202-050-0
:GHS07
Storage Temperature:Ambient