Order Entry
Germany
ContactUsLinkComponent
1,2,3,4-Tetrahydroisoquinoline 95%
1,2,3,4-Tetrahydroisoquinoline 95%
Catalog # ACRO138200250
CAS Number:  
undefined
1,2,3,4-Tetrahydroisoquinoline 95%
Catalog # ACRO138200250
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    Clear yellow to brown Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5665 to 1.5685 (20°C, 589 nm)
  • GC:
    ≥94 %

Specification Test Results

About this item

 :
 :133.19 g/mol
Boiling Pt:232…233 °C (760 mmHg)
Melting Pt:–30 °C
Density:1.06 g/cm³
Flash Pt:98 °C
Storage Temperature:Ambient
EINECS:202-050-0
UN:2735
ADR:8,II
 :GHS07
 :GHS05
CAS Number:91-21-4
Formula:C₉H₁₁N
MDL Number:MFCD00006896