Order Entry
Germany
ContactUsLinkComponent
 
Gadolinium standard solution, 1000 mg/L Gd in 0,5 M nitric acid (de Gd?O?) standard for ICP
  KRAF03720.0000
 :  
Gadolinium standard solution, 1000 mg/L Gd in 0,5 M nitric acid (de Gd?O?) standard for ICP
  KRAF03720.0000
 :  

 

  • Concentration:
    1000 mg/L
  • Matrix:
    0,5 M HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Element:
    Gadolinium
  • Symbol:
    Gd

 

 

 : Traceable to NIST