

Isobutylene oxide (2,2-dimethyloxirane, 1,2-epoxy-2-methylpropane) 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:558-30-5
:
Formula:C₄H₈O
MDL Number:MFCD00066354
:72.11 g/mol
Flash Pt:–32 °C
Storage Temperature:Ambient
EINECS:209-193-8
UN:2924
ADR:3,II
:GHS05
:GHS02
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.3705 to 1.3745 (20°C, 589 nm)
- GC:≥97.5 %