Sodium standard solution, 1,000 mg/L Na in water (from NaCl) standard for AAS
Catalog # KRAF03183.2000
Supplier: Bernd Kraft
CAS Number:
Specifications
- Concentration:1000 mg/L
- Matrix:H₂O
- Pack type:Plastic bottle
- Pk:500 mL
- Element:Sodium
- Symbol:Na
Specifications
About this item
Certifications: Traceable to NIST