Lutetium standard solution, 1000 mg/L Lu in 0,5 M nitric acid (from Lu₂O₃) standard for ICP
KRAF03775.0000
:
- Concentration:1000 mg/L
- Matrix:0,5 M HNO₃
- Pack type:Plastic bottle
- Pk:100 mL
- Element:Lutetium
- Symbol:Lu
Traceable to NIST
: Traceable to NIST