Order Entry
Germany
ContactUsLinkComponent
 
Potassium standard solution, 10000 mg/L K in 5% nitric acid (from K) standard for AAS and ICP
  634-0202
 :  
Potassium standard solution, 10000 mg/L K in 5% nitric acid (from K) standard for AAS and ICP
  634-0202
 :  

 

  • Concentration:
    10000mg/L
  • Matrix:
    5% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    500 mL
  • Element:
    Potassium
  • Symbol:
    K

 

 

 : Traceable to SRM from NIST