Vanadium standard solution, 1000 mg/L V in 1 M nitric acid (from (NH₄)VO₃) standard for AAS
Catalog # KRAF03384.2000
Supplier: Bernd Kraft
CAS Number:
Specifications
- Concentration:1000 mg/L
- Matrix:1 M HNO₃
- Pack type:Plastic bottle
- Pk:500 mL
- Element:Vanadium
- Symbol:V
Specifications
About this item
Certifications: Traceable to NIST