Mercury standard solution, 1000 mg/L Hg in 0,5 M nitric acid (from HgNO₃.1H₂O) standard for AAS
Catalog # KRAF03344.2000
Supplier: Bernd Kraft
CAS Number:
Specifications
- Concentration:1000 mg/L
- Matrix:0,5 M HNO₃
- Pack type:Plastic bottle
- Pk:500 mL
- Element:Mercury
- Symbol:Hg
Specifications
About this item
Certifications: Traceable to NIST