Mercury standard solution, 10 mg/L Hg in 5% nitric acid (from Hg) standard for ICP-MS
Catalog # JOBICLHG2-1AY
Supplier: JOBIN YVON EMISSION
CAS Number:
Specifications
- Concentration:10 mg/L
- Matrix:5% HNO₃
- Pack type:Glass bottle
- Pk:125 mL
- Element:Mercury
- Symbol:Hg
Specifications
About this item
Certifications: Traceable to SRM from NIST