Molybdenum standard solution, 1000 mg/L Mo in water 0,5 M ammonium hydroxide (from (NH₄)₆Mo₇O₂₄.4H₂O) standard for AAS
Catalog # KRAF03343.2000
Supplier: Bernd Kraft
CAS Number:
Specifications
- Concentration:1000 mg/L
- Matrix:H₂O
- Pack type:Plastic bottle
- Pk:500 mL
- Element:Molybdenum
- Symbol:Mo
Specifications
About this item
Certifications: Traceable to NIST