Gold standard solution, 1000 mg/L Au in 1 M hydrochloric acid (from HAuCl?) standard for ICP
Catalog # KRAF03472.0000
Supplier: Bernd Kraft
CAS Number:
Specifications
- Concentration:1000 mg/L
- Matrix:1 M HCl
- Pack type:Plastic bottle
- Pk:100 mL
- Element:Gold
- Symbol:Au
Specifications
About this item
Certifications: Traceable to NIST