Order Entry
Germany
ContactUsLinkComponent
Magnesium standard solution, 10000 mg/L Mg in 5% nitric acid (from Mg) standard for AAS and ICP
Magnesium standard solution, 10000 mg/L Mg in 5% nitric acid (from Mg) standard for AAS and ICP
Catalog # JOBIPLMG2-3Y
CAS Number:  
Magnesium standard solution, 10000 mg/L Mg in 5% nitric acid (from Mg) standard for AAS and ICP
Catalog # JOBIPLMG2-3Y
CAS Number:  

Specifications

  • Concentration:
    10000 mg/L
  • Matrix:
    5% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    125 mL
  • Element:
    Magnesium
  • Symbol:
    Mg

Specifications

About this item

Certifications: Traceable to SRM from NIST