Sulphuric acid 0.05 mol/l (0.1 N) in aqueous solution volumetric solution
CHMP815749501.1000
: CHEMPUR
:
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
:
:98.08 g/mol
:1.01 g/cm³ (20 °C)
:Ambient
:231-639-5
:2796
:8,II
:7664-93-9
:H₂SO₄
:MFCD00064589