Tin standard solution, 1000 mg/L Sn in 20% hydrochloric acid standard for ICP
Catalog # JOBIJYICP-SN
Supplier: JOBIN YVON EMISSION
CAS Number:
Specifications
- Concentration:1000 mg/L
- Matrix:20% HCl
- Pack type:Glass bottle
- Pk:100 mL
- Element:Tin
- Symbol:Sn