Titanium standard solution, 1,000 mg/l Ti in water with hydrofluoric acid (max. 1%) standard for ICP
Catalog # JOBIJYICP-TI
Supplier: JOBIN YVON EMISSION
CAS Number:
Specifications
- Concentration:1000 mg/L
- Matrix:H₂O/tr HF
- Pack type:Glass bottle
- Pk:100 mL
- Element:Titanium
- Symbol:Ti