Phosphorus standard solution, 1000 mg/L P in water (from H₃PO₄) standard for AAS
Catalog # KRAF11465.2000
Supplier: Bernd Kraft
CAS Number:
Specifications
- Concentration:1000 mg/L
- Matrix:H₂O
- Pack type:Plastic bottle
- Pk:500 mL
- Element:Phosphorus
- Symbol:P
Specifications
About this item
Certifications: Traceable to NIST