Phosphorus standard solution, 1000 mg/L P in water (from KH₂PO₄) standard for ICP
Catalog # KRAF10882.0000
Supplier: Bernd Kraft
CAS Number:
Specifications
- Concentration:1000 mg/L
- Matrix:H₂O
- Pack type:Plastic bottle
- Pk:100 mL
- Element:Phosphorus
- Symbol:P
Specifications
About this item
Certifications: Traceable to NIST