Copper standard solution, 1000 mg/L Cu in dil. nitric acid (from Cu) standard for AAS and ICP
Catalog # JOBIPLCU2-2Y
Supplier: JOBIN YVON EMISSION
CAS Number:
Specifications
- Concentration:1000 mg/L
- Matrix:Nitric acid dil.
- Pack type:Plastic bottle
- Pk:125 mL
- Element:Copper
- Symbol:Cu
Specifications
About this item
Certifications: Traceable to SRM from NIST