Copper standard solution, 1000 mg/L Cu in 0,5 M nitric acid (from Cu) standard for ICP
Catalog # KRAF03713.0000
Supplier: Bernd Kraft
CAS Number:
Specifications
- Concentration:1000 mg/L
- Matrix:0,5 M HNO₃
- Pack type:Plastic bottle
- Pk:100 mL
- Element:Copper
- Symbol:Cu
Specifications
About this item
Certifications: Traceable to NIST