Potassium standard solution, 1000 mg/L K in dilute nitric acid (from K) standard for ICP-MS
Catalog # JOBICLK2-2Y
Supplier: JOBIN YVON EMISSION
CAS Number:
Specifications
- Concentration:1000 mg/L
- Matrix:Nitric acid dil.
- Pack type:Plastic bottle
- Pk:100 mL
- Element:Potassium
- Symbol:K
Specifications
About this item
Certifications: Traceable to SRM from NIST