Calcium standard solution, 1000 mg/L Ca in dil. nitric acid (from Ca) standard for ICP-MS
JOBICLCA2-2Y
:
- Concentration:1000 mg/L
- Matrix:Nitric acid dil.
- Pack type:Plastic bottle
- Pk:100 mL
- Element:Calcium
- Symbol:Ca
: Traceable to SRM from NIST