Platinum(II) acetylacetonate ≥98%
Catalog # STRE78-1400-5G
Supplier: Strem Chemicals, Inc.
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:393.3 g/mol
Melting Pt:250…252 °C
Density:∼1 g/cm³ (20 °C)
EINECS:239-223-5
:GHS07
CAS Number:15170-57-7
Formula:C₁₀H₁₄O₄Pt
MDL Number:MFCD00000028