Zirconium standard solution, 1000 mg/L Zr in 2 M hydrochloric acid (from ZrOCl₂.8H₂O) standard for AAS
KRAF03478.2000
:
- Concentration:1000 mg/L
- Matrix:2 M HCl
- Pack type:Plastic bottle
- Pk:500 mL
- Element:Zirconium
- Symbol:Zr
: Traceable to NIST