Order Entry
Germany
ContactUsLinkComponent
Zinc standard solution, 1000 mg/L Zn in dil. nitric acid (from Zn) standard for AAS and ICP
Zinc standard solution, 1000 mg/L Zn in dil. nitric acid (from Zn) standard for AAS and ICP
  JOBIPLZN2-2Y
 :  
Zinc standard solution, 1000 mg/L Zn in dil. nitric acid (from Zn) standard for AAS and ICP
  JOBIPLZN2-2Y
 :  

 

  • Concentration:
    1000 mg/L
  • Matrix:
    Nitric acid dil.
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Element:
    Zinc
  • Symbol:
    Zn

 

 

 : Traceable to SRM from NIST