Order Entry
Germany
ContactUsLinkComponent
 
Tungsten standard solution, 1000 mg/L W in water (from (NH₄)₂WO₄) standard for AAS
  KRAF21563.2000
 :  
Tungsten standard solution, 1000 mg/L W in water (from (NH₄)₂WO₄) standard for AAS
  KRAF21563.2000
 :  

 

  • Concentration:
    1000 mg/L
  • Matrix:
    H₂O
  • Pack type:
    Plastic bottle
  • Pk:
    500 mL
  • Element:
    Tungsten
  • Symbol:
    W

 

 

 : Traceable to NIST