Titanium standard solution, 1000 mg/L Ti in 2 M hydrochloric acid (from TiCl₄) standard for AAS
Catalog # KRAF03641.2000
Supplier: Bernd Kraft
CAS Number:
Specifications
- Concentration:1000 mg/L
- Matrix:2 M HCl
- Pack type:Plastic bottle
- Pk:500 mL
- Element:Titanium
- Symbol:Ti
Specifications
About this item
Certifications: Traceable to NIST