Platinum standard solution, 1,000 mg/L Pt in 10% hydrochloric acid standard for ICP
Catalog # JOBIJYICP-PT
Supplier: JOBIN YVON EMISSION
CAS Number:
Specifications
- Concentration:1000 mg/L
- Matrix:10% HCl
- Pack type:Glass bottle
- Pk:100 mL
- Element:Platinum
- Symbol:Pt