Order Entry
Germany
ContactUsLinkComponent
 
Niobium standard solution, 1000 mg/L Nb in water with hydrofluoric acid (max. 1%) standard for ICP
Catalog # JOBIJYICP-NB
CAS Number:  
Niobium standard solution, 1000 mg/L Nb in water with hydrofluoric acid (max. 1%) standard for ICP
Catalog # JOBIJYICP-NB
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    H₂O/tr HF
  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Element:
    Niobium
  • Symbol:
    Nb

Specifications

About this item