Order Entry
Germany
ContactUsLinkComponent
Nickel standard solution, 1000 mg/L Ni in dil. nitric acid (from Ni) standard for AAS and ICP
Nickel standard solution, 1000 mg/L Ni in dil. nitric acid (from Ni) standard for AAS and ICP
Catalog # JOBIPLNI2-2Y
CAS Number:  
Nickel standard solution, 1000 mg/L Ni in dil. nitric acid (from Ni) standard for AAS and ICP
Catalog # JOBIPLNI2-2Y
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    Nitric acid dil.
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Element:
    Nickel
  • Symbol:
    Ni

Specifications

About this item

Certifications: Traceable to SRM from NIST