Order Entry
Germany
ContactUsLinkComponent
 
Gold standard solution, 1000 mg/L Au in 10% hydrochloric acid standard for ICP
  JOBIJYICP-AU
 :  
Gold standard solution, 1000 mg/L Au in 10% hydrochloric acid standard for ICP
  JOBIJYICP-AU
 :  

 

  • Concentration:
    1000 mg/L
  • Matrix:
    10% HCl
  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Element:
    Gold
  • Symbol:
    Au