Order Entry
Germany
ContactUsLinkComponent
Cobalt standard solution, 1,000 mg/L Co in dil. nitric acid standard for ICP
Cobalt standard solution, 1,000 mg/L Co in dil. nitric acid standard for ICP
Catalog # JOBIJYICP-CO
CAS Number:  
Cobalt standard solution, 1,000 mg/L Co in dil. nitric acid standard for ICP
Catalog # JOBIJYICP-CO
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    Nitric acid dil.
  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Element:
    Cobalt
  • Symbol:
    Co

Specifications

About this item