Order Entry
Germany
ContactUsLinkComponent
 
Cobalt standard solution, 1000 mg/L Co in 0,5 M nitric acid (from Co(NO₃)₂.6H₂O) standard for AAS
  KRAF03647.2000
 :  
Cobalt standard solution, 1000 mg/L Co in 0,5 M nitric acid (from Co(NO₃)₂.6H₂O) standard for AAS
  KRAF03647.2000
 :  

 

  • Concentration:
    1000 mg/L
  • Matrix:
    0,5 M HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    500 mL
  • Element:
    Cobalt
  • Symbol:
    Co

 

 

 : Traceable to NIST