Sulphur standard solution, 10000 mg/L S in water (from (NH₄)₂SO₄) standard for AAS
Catalog # KRAF04585.0000
Supplier: Bernd Kraft
CAS Number:
Specifications
- Concentration:10000 mg/L
- Matrix:H₂O
- Pack type:Plastic bottle
- Pk:100 mL
- Element:Sulphur
- Symbol:S