Order Entry
Germany
ContactUsLinkComponent
 
Sulphur standard solution, 1000 mg/L S in water standard for ICP
  JOBIJYICP-S
 :  
Sulphur standard solution, 1000 mg/L S in water standard for ICP
  JOBIJYICP-S
 :  

 

  • Concentration:
    1000 mg/L
  • Matrix:
    H₂O
  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Element:
    Sulphur
  • Symbol:
    S