Order Entry
Germany
ContactUsLinkComponent
 
Sulphur standard solution, 1000 mg/L S in water (from (NH₄)₂SO₄) standard for AAS
Catalog # KRAF03939.2000
Supplier:  Bernd Kraft
CAS Number:  
Sulphur standard solution, 1000 mg/L S in water (from (NH₄)₂SO₄) standard for AAS
Catalog # KRAF03939.2000
Supplier:  Bernd Kraft
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    H₂O
  • Pack type:
    Plastic bottle
  • Pk:
    500 mL
  • Element:
    Sulphur
  • Symbol:
    S

Specifications

About this item

Certifications: Traceable to NIST