Fluoride standard solution, 1000 mg/L F- in Water (from NaF) standard for ion chromatography
Supplier: Bernd Kraft
Certificates
Fluoride standard solution, 1000 mg/L F- in Water (from NaF) standard for ion chromatography
Supplier: Bernd Kraft
About this item
Certifications: Traceable to SRM from NIST
Frequently Bought Together
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Pasteur pipettes, disposable
Supplier: Brand
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Nitric acid 65%, Suprapur® for trace analysis, Supelco®
Supplier: Merck
Sodium salicylate, EMSURE® for analysis, Supelco®
Supplier: Merck
VWR®, Nitrile Gloves
Supplier: VWR Collection
Chromate standard
Supplier: Bernd Kraft
Phosphorus standard solution, 1000 mg/L P in water (from KH₂PO₄) standard for ICP
Supplier: Bernd Kraft