Order Entry
Germany
ContactUsLinkComponent
Hydrochloric acid 3 mol/l (3 N) volumetric solution
 
Hydrochloric acid 3 mol/l (3 N) volumetric solution
  CHMP805313167.1000
 :  CHEMPUR
 :  
 
Hydrochloric acid 3 mol/l (3 N) volumetric solution
  CHMP805313167.1000
 :  CHEMPUR
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

 

 

 :
 :1.06 g/cm³ (20 °C)
 :Ambient
 :231-595-7
 :8,II
 :36.46 g/mol
 :1789
 :GHS07
 :7647-01-0
 :HCl
 :MFCD00011324