Order Entry
Germany
ContactUsLinkComponent
 
Silver standard solution, 1,000 mg/L Ag in 0,5 M nitric acid (from AgNO₃) standard for AAS
  KRAF03499.2700
 :  
Silver standard solution, 1,000 mg/L Ag in 0,5 M nitric acid (from AgNO₃) standard for AAS
  KRAF03499.2700
 :  

 

  • Concentration:
    1000 mg/L
  • Matrix:
    0,5 M HNO₃
  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Element:
    Silver
  • Symbol:
    Ag

 

 

 : Traceable to NIST