Magnesium standard solution, 1000 mg/L Mg in dil. nitric acid (from Mg) standard for AAS and ICP
JOBIPLMG2-2Y
:
- Concentration:1000 mg/L
- Matrix:Nitric acid dil.
- Pack type:Plastic bottle
- Pk:125 mL
- Element:Magnesium
- Symbol:Mg
: Traceable to SRM from NIST