Order Entry
Germany
ContactUsLinkComponent
 
Chromium standard solution, 1,000 mg/L Cr in dil. nitric acid standard for ICP
  JOBIJYICP-CR
 :  
Chromium standard solution, 1,000 mg/L Cr in dil. nitric acid standard for ICP
  JOBIJYICP-CR
 :  

 

  • Concentration:
    1000 mg/L
  • Matrix:
    Nitric acid dil.
  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Element:
    Chromium
  • Symbol:
    Cr