Order Entry
Germany
ContactUsLinkComponent
 
Chromium standard solution, 1000 mg/L Cr in 0,5 M nitric acid (from CrO₃) standard for ICP
  KRAF03836.0000
 :  
Chromium standard solution, 1000 mg/L Cr in 0,5 M nitric acid (from CrO₃) standard for ICP
  KRAF03836.0000
 :  

 

  • Concentration:
    1000 mg/L
  • Matrix:
    0,5 M HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Element:
    Chromium
  • Symbol:
    Cr

 

 

 : Traceable to NIST